missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Antibody (OTI3F3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73301CL1
This item is not returnable.
View return policy
Description
PDF Monoclonal antibody specifically detects PDF in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
| Full length human recombinant protein of human PDF (NP_071736) produced in E.coli. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction | |
| 64146 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI3F3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction