missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE9A, Mouse anti-Human, Clone: 1E1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005152-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: EGLPVAPFMDRDKVTKATAQIGFIKFVLIPMFETVTKLFPMVEEIMLQPLWESRDRYEELKRIDDAMKELQKKTDSLTSGATEKSRERSRDVKNSEGDCASpecifications
| PDE9A | |
| Monoclonal | |
| Unconjugated | |
| phosphodiesterase 9A | |
| HSPDE9A2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1E1 | |
| In 1x PBS, pH 7.4 | |
| NM_001001567 | |
| PDE9A | |
| PDE9A (NP_001001567.1, 434 a.a. ∼ 533 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5152 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction