missing translation for 'onlineSavingsMsg'
Learn More

PDE9A Antibody, Novus Biologicals™

Product Code. 18325723 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325723 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325723 Supplier Novus Biologicals Supplier No. NBP312248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE9A Polyclonal antibody specifically detects PDE9A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE9A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:250
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A
Host Species Rabbit
Immunogen Synthetic cyclic peptide common to all PDE9A variants
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5152
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.