missing translation for 'onlineSavingsMsg'
Learn More

PDE9A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18417749 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18417749 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417749 Supplier Novus Biologicals Supplier No. NBP299870MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE9A Polyclonal antibody specifically detects PDE9A in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Classification Polyclonal
Purification Method Antigen and protein A Affinity-purified
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Content And Storage Store at 4C in the dark.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.