missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDE9A Polyclonal specifically detects PDE9A in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | PDE9A |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant PDE9A (Pro181-Lys506). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?