missing translation for 'onlineSavingsMsg'
Learn More

PDE8B Antibody, DyLight 350, Novus Biologicals™

Product Code. 18401173 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401173 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401173 Supplier Novus Biologicals Supplier No. NBP171817UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE8B Polyclonal antibody specifically detects PDE8B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE8B
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 3'-5' cyclic nucleotide phosphodiesterase 8B, ADSD, Cell proliferation-inducing gene 22 protein, EC 3.1.4.17, FLJ11212, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8B, hsPDE8B, phosphodiesterase 8B, PPNAD3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 835 and 885 of human Phosphodiesterase 8B using the numbering given in entry NP_003710.1 (GeneID 8622).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8622
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.