missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE8A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35230-100ul
This item is not returnable.
View return policy
Description
PDE8A Polyclonal antibody specifically detects PDE8A in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PDE8A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human PDE8A (NP_002596.1).,, Sequence:, MGCAPSIHISERLVAEDAPSPAAPPLSSGGPRLPQGQKTAALPRTRGAGLLESELRDGSGKKVAVADVQFGPMRFHQDQLQVLLVFTKEDNQCNGFCRACEKAGFKCTVTKEAQAVLACF | |
| 100 μL | |
| Lipid and Metabolism, Signal Transduction | |
| 5151 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction