missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE8A Antibody (EM-52), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62213AF750
This item is not returnable.
View return policy
Description
PDE8A Monoclonal specifically detects PDE8A in Human samples. It is validated for Western Blot.
Specifications
| PDE8A | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDE8A | |
| Human PDE8a | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-115 recognizes an epitope in the D1 domain of CD4 antigen, a 55 kDa transmebrane glycoprotein expressed on a subset of T lymphocytes and also on monocytes, tissue macrophages and granulocytes. It is negative in Western blotting even with non-reduced samples of cell lysates. | |
| Store at 4°C in the dark. |
| Western Blot | |
| EM-52 | |
| Western Blot | |
| cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A | |
| Mouse | |
| Protein A purified | |
| Lipid and Metabolism, Signal Transduction | |
| 5151 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction