missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE6H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583757
This item is not returnable.
View return policy
Description
Immunogen sequence: PRKGPPKFKQ RQTRQFKSKP PKKGVKGFGD DIPGMEGLGT DITVICPWEA FSHLELHELA Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Participates in processes of transmission and amplification of the visual signal. cGMP-PDEs are the effector molecules in G-protein-mediated phototransduction in vertebrate rods and cones.
Specifications
| PDE6H | |
| Polyclonal | |
| Unconjugated | |
| PDE6H | |
| A930033D18Rik; ACHM6; cone phosphodiesterase 6 gamma; cone-like cGMP-phosphodiesterase 6 gamma subunit type 2; GMP-PDE gamma; Pde6g; Pde6h; PDEgamma; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase 6H; phosphodiesterase 6H, cGMP-specific, cone, gamma; RCD3; retinal cone rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114248, 5149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61250, Q13956 | |
| PDE6H | |
| Recombinant protein corresponding to Human PDE6H. Recombinant protein control fragment (Product #RP-108364). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction