Learn More
PDE6B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22063-1-AP-150UL
 View more versions of this product
Product Code. 16891605
  • £349.00 / 150µL
Description
The second messengers cAMP and cGMP are key regulatory molecules that are involved in a wide variety of signal transduction pathways. Levels of cAMP and cGMP are regulated by their rate of synthesis by nucleotide cyclases and by their rate of hydrolysis by cyclic nucleotide phosphodiesterases (PDEs). PDEs form a superfamily of enzymes that catalyze the conversion of 3-prime, 5-prime-cyclic nucleotides to the corresponding nucleoside 5-prime-monophosphates. PDE6 is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha′ (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). The catalytic core of the PDE6 system is comprised of alpha′/alpha′ homodimers in the cone and alpha/beta heterodimers in the rod. The C-terminus of both the catalytic and inhibitory subunits is modified by methylation, myristyolation and prenylation which have been shown to be critical for proper complex assembly and membrane association.
Specifications
Specifications
PDE6B
Polyclonal
Unconjugated
PDE6B
CSNB3, GMP PDE beta, PDE6B, PDEB, rd1, RP40
Rabbit
Antigen Affinity Chromatography
RUO
18587, 5158
-20°C
Liquid
Immunoprecipitation, Western Blot
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23440, P35913
PDE6B
PDE6B Fusion Protein Ag17139
150 μL
Primary
Human, Mouse