Learn More
PDE6A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1721
Description
PA1-721 detects phosphodiesterase 6 alpha’ (PDE6 alpha’) from sheep, human and bovine retinal extract. PA1-721 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽90 kDa protein representing PDE6 alpha’ from sheep and bovine retinal extract. The PA1-721 immunogen is a synthetic peptide corresponding to residues M(16) P Q F A K E Y F D R K L R(29) C of human PDE6 alpha’. This sequence is completely conserved between human, chicken and frog. PA1-721 immunizing peptide (Cat. No. PEP-149) is available for use in neutralization and control experiments.
This gene encodes the cyclic-GMP (cGMP)-specific phosphodiesterase 6A alpha subunit, expressed in cells of the retinal rod outer segment. The phosphodiesterase 6 holoenzyme is a heterotrimer composed of an alpha, beta, and two gamma subunits. cGMP is an important regulator of rod cell membrane current, and its dynamic concentration is established by phosphodiesterase 6A cGMP hydrolysis and guanylate cyclase cGMP synthesis. The protein is a subunit of a key phototransduction enzyme and participates in processes of transmission and amplification of the visual signal. Mutations in this gene have been identified as one cause of autosomal recessive retinitis pigmentosa.
Specifications
| PDE6A | |
| Polyclonal | |
| PDE6A | |
| Phosphodiesterase 6 Alpha′ | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 281973, 5145, 101122024 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| P11541, P16499 | |
| PDE6A | |
| Synthetic peptide corresponding to residues M(16) P Q F A K E Y F D R K L R(29) C of human PDE6 alpha’. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Ovine | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.