missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE5A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22624-1-AP-150UL
This item is not returnable.
View return policy
Description
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.Specifications
| PDE5A | |
| Polyclonal | |
| Unconjugated | |
| PDE5A | |
| CGB PDE, CN5A, PDE5, PDE5A, PDE5A1, phosphodiesterase 5A, cGMP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 242202, 8654 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O76074, Q8CG03 | |
| PDE5A | |
| PDE5A Fusion Protein Ag18779 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction