Learn More
Invitrogen™ PDE5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550727
Product Code. 15654655
  • £443.00 / 100µL
Description
The antibody detects endogenous levels of total PDE5A protein.
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDE5
Polyclonal
Unconjugated
Pde5a
Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5
Rabbit
Antigen affinity chromatography
RUO
8654
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O76074
Pde5a
Synthetic peptide corresponding to a region derived from internal residues of human phosphodiesterase 5A, cGMP-specific.