missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDE5A Polyclonal antibody specifically detects PDE5A in Human, Mouse, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot, Immunofluorescence, Immunocytochemistry
Specifications
Specifications
| Antigen | PDE5A |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Dot Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunoprecipitation 1:200, Dot Blot 1:10000, Immunofluorescence 1:100, Immunocytochemistry 1:100 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | CGB-PDE, cGMP-binding cGMP-specific 3'-5'-cyclic nucleotide phosphodiesterase, cGMP-binding cGMP-specific phosphodiesterase, cGMP-specific 3'-5'-cyclic phosphodiesterase, cGMP-specific phosphodiesterase type 5A, CN5A, EC 3.1.4, EC 3.1.4.35, PDE5, phosphodiesterase 5A, cGMP-specific, phosphodiesterase isozyme 5 |
| Host Species | Rabbit |
| Immunogen | Phosphorylated synthetic peptide common to all PDE5A variants taken within amino acid region 120-170 on human PDE5A protein. |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?