Learn More
Invitrogen™ PDE5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117072
Description
Positive Samples: HT-1080, SKOV3, A-549, 22Rv1, Mouse brain, Mouse skeletal muscle, Rat skeletal muscle Immunogen sequence: MERAGPSFGQ QRQQQQPQQQ KQQQRDQDSV EAWLDDHWDF TFSYFVRKAT REMVNAWFAE RVHTIPVCKE GIRGHTESCS CPLQQSPRAD NSAPGTPTRK ISASEFDRPL RPIVVKDSEG TVSFLSDSEK KEQMPLTPPR FDHDEGDQCS.
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
Specifications
| PDE5 | |
| Polyclonal | |
| Unconjugated | |
| Pde5a | |
| Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171115, 242202, 8654 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54735, O76074, Q8CG03 | |
| Pde5a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human PDE5A (NP_0010742). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.