Learn More
Invitrogen™ PDE5 Monoclonal Antibody (K1E005_9H12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556207
Description
Sequence of this protein is as follows: QSLAAAVVPS AQTLKITDFS FSDFELSDLE TALCTIRMFT DLNLVQNFQM KHEVLCRWIL SVKKNYRKNV AYHNWRHAFN TAQCMFAALK AGKIQNKLTD LEILALLIAA LSHDLDHRGV NNSYIQRSEH PLAQLYCHSI MEHHHFDQCL MILNSPGNQI LSGLSIEEYK TTLKIIKQAI LATDLALYIK RRGEFFELIR KNQFNLEDPH QKELFLAMLM TACDLSAITK PWPIQQRIAE LVATEFFDQG DRERKELNIE PTDLMNREKK NKIPSMQVGF IDAICLQLYE ALTHVSEDCF PLLDGCRKNR QKWQALAEQQ
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
Specifications
| PDE5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O76074 | |
| Pde5a | |
| Recombinant human PDE5A (immunogen range is 541-860). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K1E005_9H12 | |
| Unconjugated | |
| Pde5a | |
| Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 8654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.