Learn More
Invitrogen™ PDE5 Monoclonal Antibody (K1E005_13D12), Invitrogen™
Mouse Monoclonal Antibody
£453.00 - £760.00
Specifications
| Antigen | PDE5 |
|---|---|
| Clone | K1E005_13D12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210785
|
Invitrogen™
MA556319 |
200 μL |
£760.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30194909
|
Invitrogen™
MA555939 |
50 μL |
£453.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: QSLAAAVVPS AQTLKITDFS FSDFELSDLE TALCTIRMFT DLNLVQNFQM KHEVLCRWIL SVKKNYRKNV AYHNWRHAFN TAQCMFAALK AGKIQNKLTD LEILALLIAA LSHDLDHRGV NNSYIQRSEH PLAQLYCHSI MEHHHFDQCL MILNSPGNQI LSGLSIEEYK TTLKIIKQAI LATDLALYIK RRGEFFELIR KNQFNLEDPH QKELFLAMLM TACDLSAITK PWPIQQRIAE LVATEFFDQG DRERKELNIE PTDLMNREKK NKIPSMQVGF IDAICLQLYE ALTHVSEDCF PLLDGCRKNR QKWQALAEQQ
PDE5 Monoclonal antibody specifically detects PDE5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| PDE5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5 | |
| Recombinant human PDE5A (immunogen range is 541-860). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Pde5a |
| K1E005_13D12 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O76074 | |
| 8654 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.