missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE4D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12918-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of four mammalian counterparts to the fruit fly ′dunce′ gene. The encoded protein has 3′,5′-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.Specifications
| PDE4D | |
| Polyclonal | |
| Unconjugated | |
| PDE4D | |
| DPDE3, HSPDE4D, PDE43, PDE4D, PDE4DN2, STRK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 238871, 24627, 5144 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14270, Q01063, Q08499 | |
| PDE4D | |
| PDE4D Fusion Protein Ag3588 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction