Learn More
Invitrogen™ PDE4D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589542
Description
Immunogen sequence: FKLLQEENCD IFQNLTKKQR QSLRKMVIDI VLATDMSKHM NLLADLKTMV ETKKVTSSGV LLLDNYSDRI QVLQNMVHCA DLSNPTKPLQ LYRQWTDRIM EEFFRQGDRE RERGMEISPM CDKHNASVEK SQVGFIDYIV HPLWETWADL VHPDAQDILD TLEDNREWYQ STIPQSPSPA PDDPEEGRQG QTEKFQFELT LEEDGESDTE KDSGSQVEED TSCSDSKTLC TQDSESTEIP LDEQVEEEAV GEEEESQPEA CVIDDRSPDT; Positive Samples: HT-29, A-549, SW480; Cellular Location: Apical cell membrane, Cytoplasm, Membrane, centrosome, cytoskeleton, microtubule organizing center.
This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.
Specifications
| PDE4D | |
| Polyclonal | |
| Unconjugated | |
| Pde4d | |
| 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24627, 5144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14270, Q08499 | |
| Pde4d | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 249-518 of human PDE4D (NP_0011841521). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.