Learn More
Invitrogen™ PDE4D Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA131136
Product Code. 11511623
  • £552.00 / 100µg
Description
PA1-31136 detects PDE4D from mouse, human, rat samples.
This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDE4D
Polyclonal
Unconjugated
Pde4d
9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136
Rabbit
Affinity chromatography
RUO
238871, 24627, 5144
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
0.78 mg/mL
Tris glycine with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-4.9
P14270, Q01063, Q08499
Pde4d
C-terminal cyclic peptide unique to all PDED subtypes variants.
100 μg
Primary
Human, Mouse, Rat