missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE4B Antibody (OTI1D12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73292DL594
This item is not returnable.
View return policy
Description
PDE4B Monoclonal antibody specifically detects PDE4B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PDE4B | |
| Monoclonal | |
| DyLight 594 | |
| cAMP-specific (phosphodiesterase E4 dunce homolog, cAMP-specific 3'-5'-cyclic phosphodiesterase 4B, cAMP-specific phosphodiesterase-4 B isoform, DKFZp686F2182, Drosophila), dunce-like phosphodiesterase E4, EC 3.1.4, EC 3.1.4.17, MGC126529, PDE32, PDEIVB, phosphodiesterase 4B, cAMP-specific, phosphodiesterase 4B, cAMP-specific (dunce, phosphodiesterase 4B, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E4), phosphodiesterase E4 dunce homolog | |
| Full length recombinant protein of human PDE4B (NP_002591) produced in HEK293T cells. | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 5142 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1D12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction