missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE3B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25290-1-AP-150UL
This item is not returnable.
View return policy
Description
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.Specifications
| PDE3B | |
| Polyclonal | |
| Unconjugated | |
| PDE3B | |
| CGI PDE B, CGIP1, cGIPDE1, HcGIP1, PDE3B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5140 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13370 | |
| PDE3B | |
| PDE3B Fusion Protein Ag18064 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction