missing translation for 'onlineSavingsMsg'
Learn More

PDE3B Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18417932 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417932 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417932 Supplier Novus Biologicals Supplier No. NBP146183AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE3B Polyclonal antibody specifically detects PDE3B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE3B
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CGIP1, CGI-PDE B, CGIPDE1, cGMP-inhibited 3'-5'-cyclic phosphodiesterase B, Cyclic GMP-inhibited phosphodiesterase B, cyclic nucleotide phosphodiesterase, EC 3.1.4, EC 3.1.4.17, HcGIP1, phosphodiesterase 3B, cGMP-inhibited
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 500 and 550 of human Phosphodiesterase 3B using the numbering given in entry NP_000913.2 (GeneID 5140).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5140
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.