missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582503
This item is not returnable.
View return policy
Description
Immunogen sequence: SPQILTPPVI CSSCGRPYSQ GNPADEPLER SGVATRTPSR TDDTAQVTSD YETNNNSDSS DIVQNEDETE CLREPLRKAS ACSTYAPETM MFLDKPILAP EPLVMDNLDS IMEQLNT Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 81%.
Cyclic nucleotide phosphodiesterases (PDEs) comprise a complex group of enzymes, and at least 5 major PDE families or classes with distinctive properties have been identified. Members of the cGMP-inhibited cAMP PDE (cGI-PDE) family, such as PDE3A, are characterized by high affinity for cAMP and cGMP and competitive inhibition of their cAMP hydrolytic activity by cGMP and certain positive inotropic agents (Meacci et al., 1992 [PubMed 1315035]).
Specifications
| PDE3A | |
| Polyclonal | |
| Unconjugated | |
| PDE3A | |
| A930022O17Rik; C87899; cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE; CGI-PDE A; CGI-PDE-A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; cGMP-inhibited phosphodiesterase; cyclic GMP-inhibited phosphodiesterase A; cyclic nucleotide phosphodiesterase; HTNB; PDE3A; phosphodiesterase 3A; phosphodiesterase 3A, cGMP inhibited; phosphodiesterase 3A, cGMP-inhibited; RNPDE3A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5139 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14432 | |
| PDE3A | |
| Recombinant protein corresponding to Human PDE3A. Recombinant protein control fragment (Product #RP-90769). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction