missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£258.00 - £410.00
Specifications
| Antigen | PDE3A |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407181
|
Novus Biologicals
NBP1-85645-25ul |
25 μL |
£258.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421482
|
Novus Biologicals
NBP1-85645 |
0.1 mL |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE3A Polyclonal antibody specifically detects PDE3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PDE3A | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 5139 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CGI-PDE, CGI-PDE A, cGMP-inhibited 3'-5'-cyclic phosphodiesterase A, cyclic GMP inhibited phosphodiesterase A, Cyclic GMP-inhibited phosphodiesterase A, EC 3.1.4, EC 3.1.4.17, phosphodiesterase 3A, cGMP-inhibited | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SPQILTPPVICSSCGRPYSQGNPADEPLERSGVATRTPSRTDDTAQVTSDYETNNNSDSSDIVQNEDETECLREPLRKASACSTYAPETMMFLDKPILAPEPLVMDNLDSIMEQLNT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title