missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE2A Rabbit anti-Human, Clone: 201, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90159-50UL
This item is not returnable.
View return policy
Description
PDE2A Monoclonal specifically detects PDE2A in Human samples. It is validated for ELISA.
Specifications
| PDE2A | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 201 | |
| ELISA 1:5000-1:10000 | |
| cGMP-dependent 3′-5′-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
| Recombinant Human PDE2A protein | |
| 50 μg | |
| Signal Transduction | |
| 5138 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction