missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1B Antibody (OTI6E4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73278CL1
This item is not returnable.
View return policy
Description
PDE1B Monoclonal antibody specifically detects PDE1B in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| PDE1B | |
| Monoclonal | |
| CoraFluor 1 | |
| 63 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B | |
| Full length human recombinant protein of human PDE1B(NP_000915) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 5153 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI6E4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction