missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97975-50ul
This item is not returnable.
View return policy
Description
PDE1B Polyclonal antibody specifically detects PDE1B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PDE1B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| 63 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B | |
| Produced in rabbits immunized with E. coli-derived Human PDE1B fragment. | |
| 50 μg | |
| Apoptosis, Signal Transduction | |
| 5153 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction