missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDE1A Polyclonal antibody specifically detects PDE1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | PDE1A |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:10000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:200 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | 3'-5' cyclic nucleotide phosphodiesterase, 61 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1A, calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, calmodulin-dependent phosphodiesterase, Cam-PDE 1A, EC 3.1.4, EC 3.1.4.17, hCam-1, HCAM1, HSPDE1A, MGC26303, phosphodiesterase 1A, calmodulin-dependent, phosphodiesterase-1A |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to unique amino acid sequence within amino acid region 1-50 on PDE1A protein. Common to all PDE1A variants. |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?