missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | PDE12 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227055
|
Novus Biologicals
NBP3-35739-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227944
|
Novus Biologicals
NBP3-35739-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE12 Polyclonal antibody specifically detects PDE12 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| PDE12 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| 2'-PDE, phosphodiesterase 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-300 of human PDE12 (NP_001309106.1).,, Sequence:, AVEKLSRAEAGSQTAAGAMERAVVRCVPSEPKLSLSFALADGSHKNMQRDQSEPLGRVLSRIATNALKGHAKAAAAKKSRKSRPNASGGAACSGPGPEPAVFCEPVVKLYYREEAVAEDVLNVDAWQDGAVLQIGDVKYKVERNPPAFTELQLPRYIMAGFPVCPKLSLEFGDPASSLFRWYKEAKPGAAEPEVGVPSSLSPSSPSSSWTETDVEERVYTPSNADIGLRLKLHCTPGDGQRFGHSRELESVCVVEAGPGTCTFDHRHLYTKKVTEDALIRTVSY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 201626 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title