missing translation for 'onlineSavingsMsg'
Learn More

PDE11A Antibody, Novus Biologicals™

Product Code. 18371962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371962 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371962 Supplier Novus Biologicals Supplier No. NBP312255

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE11A Polyclonal antibody specifically detects PDE11A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE11A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:250, Immunohistochemistry-Paraffin
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias cAMP and cGMP cyclic nucleotide phosphodiesterase 11A, cAMP and cGMP phosphodiesterase 11A, cyclic nucleotide phosphodiesterase 11A, dual 3'-5'-cyclic-AMP and -GMP phosphodiesterase 11A, EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ23693, MGC133355, MGC133356, PDE11A1, PDE11A2, PDE11A3, phosphodiesterase 11A, PPNAD2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 75-125 on PDE11A protein. Phosphorylated Ser 117.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50940
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.