missing translation for 'onlineSavingsMsg'
Learn More

PDE10-IN-1, MedChemExpress

Product Code. 16481557
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481557 50 mg 50mg
16465368 10 mg 10mg
16428087 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481557 Supplier Medchem Express Supplier No. HY12813
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PDE10-IN-1 is a potent PDE10-IN-1 inhibitor extracted from Patent WO 2013192273 A1, for treating CNS and metabolic disorders.

PDE10-IN-1 is a potent PDE10-IN-1 inhibitor extracted from Patent WO 2013192273 A1, for treating CNS and metabolic disorders.

Specifications

Chemical Name or Material PDE10-IN-1
CAS 1516896-09-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C21H19N7
Quantity 50 mg
Solubility Information DMSO : 2 mg/mL (5.41 mM; Need ultrasonic)
SMILES CC1=C([C@H]2[C@H](C3=NN4C(C(C)=NC=C4C)=N3)C2)N=C5C6=C(N=CC=C6)C=CN51
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 97.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.