missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81872
This item is not returnable.
View return policy
Description
PDCL3 Polyclonal specifically detects PDCL3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PDCL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| PDCL3 | |
| PDCL3 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human PDCL3. The immunogen is located within the last 50 amino acids of PDCL3. Amino Acid Squence: RSVLMKRDSDSEGD | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PDCL3 antibody is human, mouse and rat reactive. PDCL3 antibody is predicted to not cross-react with other members of the PDCL protein family. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin | |
| HTPHLP, IAP-associated factor VIAF1, MGC3062, PHLP3, phosducin-like 3, phosducin-like protein 3, PhPL3, VIAF, VIAF-1, VIAF1PHLP2A, Viral IAP-associated factor 1 | |
| Rabbit | |
| 25 kDa | |
| 0.1 mg | |
| Apoptosis | |
| 79031 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction