missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals PDCL Antibody (4G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00005082-M03
This item is not returnable.
View return policy
Description
PDCL Monoclonal antibody specifically detects PDCL in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| PDCL | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp564M1863, PhLP, phosducin-like, phosducin-like protein | |
| PDCL (AAH17227, 1 a.a. ~ 301 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTTLDDKLLGEKLQYYYSSSEDEDSDHEDKDRGRCAPASSSVPAEAELAGEGISVNTGPKGVINDWRRFKQLETEQREEQCREMERLIKKLSMTCRSHLDEEEEQQKQKDLQEKISGKMTLKEFAIMNEDQDDEEFLQQYRKQRMEEMRQQLHKGPQFKQVFEISSGEGFLDMIDKEQKSIVIMVHIYEDGIPGTEAMNGCMICLAAEYPAVKFCKVKSSVIGASSQFTRNALPALLIYKGGELIGNFVRVTDQLGDDFFAVDLEAFLQEFGLLPEKEVLVLTSVRNSATCHSEDSDLEID | |
| 0.1 mg | |
| Signal Transduction | |
| 5082 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 4G5 | |
| Western Blot, ELISA, Sandwich ELISA | |
| AAH17227 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction