missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDCD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588369
This item is not returnable.
View return policy
Description
Immunogen sequence: EKEKILLQKR EIESKLFGDP DEFPLAHLLE PFRQYYLQAE HSLPALIQIR HDWDQYLVPS DHPKGNFVPQ GWVLPPLPSN DIWATAVKLH; Positive Samples: HT-29, HeLa, Jurkat, MCF-7, Rat thymus; Cellular Location: Nucleus.
PDCD7 is a protein with sequence similarity to a mouse protein originally identified in embryonic stem cells. In mouse T-cell lines, this protein appears to be related to glucocorticoid- and staurine-induced apoptotic pathways, and to be linked to ceramide-mediated signaling. These observations suggest that this gene product is involved in specific apoptotic processes in T-cells. This gene encodes a protein with sequence similarity to a mouse protein originally identified in embryonic stem cells. In mouse T-cell lines, this protein appears to be related to glucocorticoid- and staurine-induced apoptotic pathways, and to be linked to ceramide-mediated signaling. These observations suggest that this gene product is involved in specific apoptotic processes in T-cells.
Specifications
| PDCD7 | |
| Polyclonal | |
| Unconjugated | |
| Pdcd7 | |
| apoptosis-related protein ES18; C80112; ES18; H MGC22015; hES18; LOW QUALITY PROTEIN: programmed cell death protein 7; MGC22015; PDCD7; programmed cell death 7; programmed cell death protein 7; Programmed cell death protein 7-like protein; U11/U12 snRNP 59K | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10081, 363082 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8N8D1 | |
| Pdcd7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 396-485 of human PDCD7 (NP_0056981). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction