missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDCD6 Recombinant Mouse Monoclonal Antibody (K92068_2D10)

Product Code. 30829621
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829621 50 μL
30825565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829621 Supplier Invitrogen™ Supplier No. MA565889

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: GAGPGPAAGA ALPDQSFLWN VFQRVDKDRS GVISDTELQQ ALSNGTWTPF NPVTVRSIIS MFDRENKAGV NFSEFTGVWK YITDWQNVFR TYDRDNSGMI DKNELKQALS GFGYRLSDQF HDILIRKFDR QGRGQIAFDD FIQGCIVLQR LTDIFRRYDT DQDGWIQVSY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%, Pig - 98%, Cynomolgus monkey - 99%.

An increased intracellular Ca2+ concentration induces apoptotic cell death. Transiently elevated Ca2+ concentrations are required for glucocorticoid-mediated and T cell receptor-mediated pathways, leading to T cell apoptosis. ALG-2 (for apoptosis-linked gene 2) is a Ca2+ -binding protein that participates in regulatory events occuring late in the apoptotic program and where several death signals converge. ALG-2 is a protein expressed in normal brain, and to a greater extent in ischemic brain. The ALG-2 protein contains five EF-hand-like motifs and shares homology with members of the penta EF-hand family, which includes Calpain small subunits sorcin and Grancalcin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD6
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K92068_2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Pdcd6
Gene Accession No. O75340
Gene Alias Alg2; alg-2; ALG-257; Apoptosis-linked gene 2 protein; Apoptosis-linked gene 2 protein homolog; AV299538; MA-3; PDCD6; PEF1B; PMP41; probable calcium-binding protein ALG-2; programmed cell death 6; programmed cell death protein 6; PS2
Gene Symbols Pdcd6
Host Species Mouse
Immunogen Recombinant human PDCD6 (aa 11-180).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10016
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.