missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527639
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (88%), Xenopus laevis (86%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
An increased intracellular Ca2+ concentration induces apoptotic cell death. Transiently elevated Ca2+ concentrations are required for glucocorticoid-mediated and T cell receptor-mediated pathways, leading to T cell apoptosis. ALG-2 (for apoptosis-linked gene 2) is a Ca2+ -binding protein that participates in regulatory events occuring late in the apoptotic program and where several death signals converge. ALG-2 is a protein expressed in normal brain, and to a greater extent in ischemic brain. The ALG-2 protein contains five EF-hand-like motifs and shares homology with members of the penta EF-hand family, which includes Calpain small subunits sorcin and Grancalcin.
Specifications
| PDCD6 | |
| Polyclonal | |
| Unconjugated | |
| Pdcd6 | |
| Alg2; alg-2; ALG-257; Apoptosis-linked gene 2 protein; Apoptosis-linked gene 2 protein homolog; AV299538; MA-3; PDCD6; PEF1B; PMP41; probable calcium-binding protein ALG-2; programmed cell death 6; programmed cell death protein 6; PS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.62 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75340 | |
| Pdcd6 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 191 of Human PDCD6 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction