missing translation for 'onlineSavingsMsg'
Learn More

PDCD6 Antibody (2B4), Novus Biologicals™

Product Code. 18341279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341279 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341279 Supplier Novus Biologicals Supplier No. H00010016M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDCD6 Monoclonal antibody specifically detects PDCD6 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD6
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2B4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12384
Gene Alias ALG2, ALG-2, Apoptosis-linked gene 2 protein, apoptosis-linked gene-2, FLJ46208, MGC111017, MGC119050, MGC9123, PEF1B, Probable calcium-binding protein ALG-2, programmed cell death 6, programmed cell death protein 6
Host Species Mouse
Immunogen PDCD6 (AAH12384, 92 a.a. ~ 191 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ITDWQNVFRTYDRDNSGMIDKNELKQALSGFGYRLSDQFHDILIRKFDRQGRGQIAFDDFIQGCIVLQRLTDIFRRYDTDQDGWIQVSYEQYLSMVFSIV
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10016
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.