missing translation for 'onlineSavingsMsg'
Learn More

PDCD5 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18408213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408213 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408213 Supplier Novus Biologicals Supplier No. NBP176991AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDCD5 Polyclonal antibody specifically detects PDCD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD5
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias FLJ42784, MGC9294, programmed cell death 5, programmed cell death protein 5, Protein TFAR19, TF-1 cell apoptosis-related protein 19, TFAR19 novel apoptosis-related, TFAR19TF1 cell apoptosis-related gene 19
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human PDCD5. The immunogen is located within the first 50 amino acids of PDCD5. Amino Acid Squence: QEAKHREAEMRNS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9141
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.