missing translation for 'onlineSavingsMsg'
Learn More

PDCD4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18324374 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324374 20 μL 20µL
18330027 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18324374 Supplier Novus Biologicals Supplier No. NBP30340120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDCD4 Polyclonal specifically detects PDCD4 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD4
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PDCD4 (NP_055271.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 27250
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.