missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4, p Ser457 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-60014
This item is not returnable.
View return policy
Description
PDCD4 Polyclonal specifically detects PDCD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDCD4 (p Ser457) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
| Rabbit | |
| 51.73 kDa | |
| 0.1 mg | |
| Apoptosis, Phospho Specific | |
| 27250 | |
| Human, Mouse, Rat, Xenopus species | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:400000, Immunohistochemistry 21.25-2.5 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q53EL6 | |
| PDCD4 | |
| This affinity purified PDCD4 [p Ser457] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids surrounding Ser457 in the human PDCD4 protein. (Uniprot: Q53EL6) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for human PDCD4 protein phosphorylated at Ser457. A BLAST analysis was used to suggest cross-reactivity with PDCD4 from human, mouse, rat and Xenopus species based on 100% homology with the immunizing sequence. Cross-reactivity with PDCD4 from other sources has not been determined. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction