missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Mouse anti-Human, Clone: 4C2, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £320.00
Specifications
| Antigen | PDCD4 |
|---|---|
| Clone | 4C2 |
| Concentration | 1 mg/ml |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18387822
|
Bio-Techne
NBP3-18224-50UL |
50 μg |
£196.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303684
|
Novus Biologicals
NBP3-18224-100UL |
100 μg |
£320.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDCD4 Monoclonal antibody specifically detects PDCD4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| PDCD4 | |
| 1 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
| Recombinant human PDCD4 (1-469aa) purified from E. coli | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 4C2 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Phospho Specific | |
| Phosphate-Buffered Saline (pH 7.4), 10% glycerol | |
| 27250 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title