missing translation for 'onlineSavingsMsg'
Learn More

PDCD4 Antibody (k4C1), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18428916 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428916 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428916 Supplier Novus Biologicals Supplier No. NBP104329JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDCD4 Monoclonal antibody specifically detects PDCD4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD4
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone k4C1
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a
Host Species Mouse
Immunogen Recombinant human PDCD4 (1-469aa) purified from E. coli (NP_055271).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 27250
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.