missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-76738-0.025mg
This item is not returnable.
View return policy
Description
PDCD4 Polyclonal specifically detects PDCD4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDCD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
| Rabbit | |
| 52 kDa | |
| 0.025 mg | |
| Apoptosis | |
| 27250 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| B5ME91 | |
| PDCD4 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human PDCD4. The immunogen is located within the last 50 amino acids of PDCD4. Amino Acid Squence: GRKRFVSEGDGGRLKPESY | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction