missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Antibody (4C2), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18224MFV450
This item is not returnable.
View return policy
Description
PDCD4 Monoclonal antibody specifically detects PDCD4 in Human samples. It is validated for Flow Cytometry
Specifications
| PDCD4 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 4C2 | |
| Flow Cytometry | |
| H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
| Recombinant human PDCD4 (1-469aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Phospho Specific | |
| 27250 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction