missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD10 Antibody (2A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43064
This item is not returnable.
View return policy
Description
PDCD10 Monoclonal antibody specifically detects PDCD10 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| PDCD10 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 2A8 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:200 | |
| apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
| programmed cell death 10 - (Uniprot# Q9BUL8) | |
| 100 μg | |
| Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology, Neuroscience, Signal Transduction | |
| 11235 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction