missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDC-TREM Rat anti-Mouse, BV650, Clone: 4A6, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 751708
This item is not returnable.
View return policy
Description
PDC-TREM Monoclonal antibody specifically detects PDC-TREM in Mouse samples. It is validated for Flow Cytometry.
Specifications
| PDC-TREM | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms PDC-TREM BV650 4A6 50 μg | |
| Mouse PDC-TREM Recombinant Protein | |
| RUO | |
| PDC-TREM | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| 4A6 | |
| Brilliant Violet 650 | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction