missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDAP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97989JF646
This item is not returnable.
View return policy
Description
PDAP1 Polyclonal specifically detects PDAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| PDAP1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 11333 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction