missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies PD98059

Product Code. 17170032
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170032 1 mg 1mg
17180032 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17170032 Supplier STEMCELL Technologies Supplier No. 72172
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MEK/ERK pathway inhibitor; Inhibits MEK1 and MEK2

  • PD98059 is a selective, cell permeable inhibitor of the MEK/ERK pathway that acts by preventing the activation of MEK1 (IC50 = 2 - 7 μM) and MEK2 (IC50 = 50 μM) by upstream kinases
  • It does not inhibit activated MEK, or the p38 MAPK pathway
  • (Alessi et al.), Davies et al.), Dudley et al.)
  • Enhances the growth and self-renewal of mouse embryonic stem (ES) cells (Burdon et al.), Qi et al.)
  • Permits derivation of mouse ES cells from the refractory CBA mouse strain (Buehr and Smith)
  • Blocks the differentiation of mouse ES cells (Burdon et al.)
  • Enhances adipogenic differentiation and blocks osteogenic differentiation of human mesenchymal stem cells (Jaiswal et al.)
  • Decreases number of AML blast colonies with minimal effect on normal hematopoietic progenitors (Milella et al.)

Specifications

CAS 167869-21-8
Quantity 1 mg
Molecular Formula C16H13NO3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.