missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PD98059

Product Code. 10314223
Click to view available options
Quantity:
5 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10314223

Brand: Invitrogen™ PHZ1164

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

PD98059 is a potent and selective cell permeable inhibitor of MAP kinase kinase (MEK). It is provided in a Lyophilized format.

PD98059 selectively blocks the activation of MEK, thereby inhibiting the phosphorylation and the activation of MAP kinase.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5604, 5605, 5606, 5607, 5608, 5609, 6416
Molecular Weight (g/mol) 267.3 Da
Purification Method HPLC
Quantity 5 mg
Sterility Non-sterile
Content And Storage Store in freezer (-5 to -30°C).
Inhibitor Properties Chemical
Inhibitors Kinase Inhibitor, Kinase Inhibitor (MAP Kinase Inhibitor)
Shipping Condition Dry Ice
Specific Inhibit or Target MEK (MAP kinase kinase)
Form Solid
Research Category Signal Transduction
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.